Human processed serum (HPS), steroid depleted
Supplier: SEQENS
Certificates
About this item
SEQENS IVD manufactures Steroids Depleted Human Processed Serum.
- Dry extract: 65 to 85 g/L
- Total proteins: 55 to 75 g/L
- Cortisol: <0,11 µg/dl
- Estradiol: <5 pg/ml
- Testosterone: <50 pg/ml
- Progesterone: <0,03 ng/ml
- Filtered through a 0,2 µm membrane
- Shelf life: 2 years
For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.
Complete donor traceability on file.
Caution: Only for research or manufacture of in vitro diagnostic reagents. Not for therapeutic use. This product of human origin should be considered as potentially infectious and handled accordingly.
Frequently Bought Together