Order Entry
Austria
ContactUsLinkComponent
Human AB pooled serum (male), French origin
Human AB pooled serum (male), French origin
Catalog # 21001PM-100ML
Supplier:  SEQENS
undefined
Human AB pooled serum (male), French origin
Catalog # 21001PM-100ML
Supplier:  SEQENS

Specifications

  • Serum format:
    Frozen
  • Description:
    Human AB pooled serum (male)
  • Pk:
    100 mL

Specifications

About this item

SEQENS IVD offers Human AB Pooled Serum (male), collected from French healthy AB male donors.

  • Dry extract: 70 to 90 g/L
  • Total proteins: 60 to 80 g/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 3 years

Each donor unit used for the manufacture of this product has been tested and found negative or non-reactive (CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.

Complete donor traceability on file.

Caution: Only for research or manufacture of in vitro diagnostic reagents. Not for therapeutic use. This product of human origin should be considered as potentially infectious and handled accordingly.