Bovine fibrinogen
Supplier: SEQENS
Certificates
About this item
SEQENS IVD provides highly purified fibrinogen manufactured according to Cohn fractionation method.
- Total proteins: >70%
- Shelf life: 3 years
- Sterile filtered 0,2 µm prior to being freeze-dried
The plasma used is collected from healthy animals of French origin. The plasma used is collected from healthy animals of French origin. Bovine Fibrinogen is developed for RPF supplement, used with Baird-Parker agar, for the detection of Staphylococcus aureus.
Full traceability and batch-to-batch repeatability guaranteed.
Delivery: Supplied lyophilised. Contains trisodium citrate and sodium chloride.
Specifications
- Species:Bovine
- Protein/peptide name:Fibrinogen
- Formulation:lyophilized
Specifications
Frequently Bought Together