Bovine serum albumin (BSA)
Supplier: SEQENS
Certificates
About this item
SEQENS IVD manufactures Bovine Serum Albumin (BSA) derived from plasma, collected from healthy animals of French origin. This product is obtained by 'Heat Shock' method, which ensures higher purity and preserves BSA intrinsic properties.
- Albumin purity: >98%
- Total proteins: >92%
- Endotoxins: <15,0 Endotoxin Unit/mg
- Shelf life: 3 years
- Sterile filtered 0,2 µm prior to being freeze-dried
Full traceability and batch-to-batch repeatability guaranteed.
Delivery: Supplied lyophilised.
Specifications
- Source:Serum
- Species:Bovine
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Endotoxin level:Low
- Formulation:lyophilized
Specifications
Frequently Bought Together