Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Anionic Standards
Sulphate Standards
Sulphate standard for ISE calibration, Orion™
Sulphate standard for ISE calibration, Orion™
Sulphate standard for ISE calibration, Orion™
662-0848
:
Thermo Orion
:
Sulphate standard for ISE calibration, Orion™
662-0848
:
Thermo Orion
:
Concentration:
0,1M
Pk:
475 mL
Description:
Sulphate standard for ISE calibration
Contents:
Na₂SO₄ in H₂O