Anti-KIF5B Goat Polyclonal Antibody (Biotinylated)
Catalog # EVBIEB05492-B
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen symbol:KIF5B
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Format:Affinity Purified
- Form:Liquid
- Gene ID:3799
- Antigen synonyms:ubiquitous kinesin heavy chain|UKHC|kinesin 1 (110-120kD)|KNS|KINH|conventional kinesin heavy chain|KNS1|kinesin family member 5B|epididymis secretory protein Li 61|kinesin heavy chain|KIF5B|HEL-S-61
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Molecular weight:110
- Sequence:CQPVAVRGGGGKQV.
- Storage temperature:+8 °C
- Concentration:0,5 µg/µl
- Immunogen:Kinesin 1 / UKHC, Biotinylated
- Size:200 μL
- Pack type:Plastic Vial
- Pk:0,1 mg
Specifications
About this item
Western Blot: Approx 60kDa band observed in Human Platelets lysates (calculated MW of 67.8kDa according to NP_009200.2). Recommended concentration: 0.1-0.3µg/ml.
Type: Primary
Antigen: KIF5B
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Cow