Order Entry
Austria
ContactUsLinkComponent
 
Anti-GPT Goat Polyclonal Antibody
Catalog # EVBIEB12973
Supplier:  Everest Biotech
Anti-GPT Goat Polyclonal Antibody
Catalog # EVBIEB12973
Supplier:  Everest Biotech

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    GPT
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Affinity Purified
  • Form:
    Liquid
  • Gene ID:
    2875
  • Antigen synonyms:
    AAT1|GPT1|GPT|glutamic--alanine transaminase 1|glutamic--pyruvic transaminase 1|glutamic-pyruvate transaminase (alanine aminotransferase)|ALT1|alanine aminotransferase 1|glutamic-alanine transaminase 1|glutamate pyruvate transaminase 1|GPT 1
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
  • Molecular weight:
    54.6
  • Sequence:
    KLTEQVFNEAPGISC
  • Storage temperature:
    +8 °C
  • Concentration:
    0,5 µg/µl
  • Immunogen:
    ALT1 / GPT1 (aa391-405)
  • Purification:
    Affinity purified
  • Size:
    200 μL
  • Pack type:
    Plastic Vial
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: GPT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human