Anti-PTGIS Goat Polyclonal Antibody
Catalog # EVBIEB12919
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen symbol:PTGIS
- Clonality:Polyclonal
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human
- Western blot:Yes
- Format:Affinity Purified
- Form:Liquid
- Gene ID:5740
- Antigen synonyms:prostaglandin I2 synthase|subfamily A|family 8|cytochrome P450|polypeptide 1|PTGIS|PTGI|prostacyclin synthase|CYP8A1|prostaglandin I2 (prostacyclin) synthase|PGIS|CYP8
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Molecular weight:57.1
- Sequence:C-RRAHRSKWLES
- Storage temperature:+8 °C
- Concentration:0,5 µg/µl
- Immunogen:PTGIS / CYP8A1 (aa248-258)
- Size:200 μL
- Pack type:Plastic Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: PTGIS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Pig