Anti-ASS1 Goat Polyclonal Antibody (Biotinylated)
Catalog # EVBIEB07697-B
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen symbol:ASS1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Format:Affinity Purified
- Form:Liquid
- Gene ID:445
- Antigen synonyms:ASS|argininosuccinate synthase 1|ASS1|citrulline-aspartate ligase|CTLN1|citrulline--aspartate ligase|argininosuccinate synthetase 1
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Molecular weight:46.5; 46.5; 50.8
- Sequence:C-ENPKNQAPPGLYTKTQD.
- Storage temperature:+8 °C
- Concentration:0,5 µg/µl
- Immunogen:Argininosuccinate synthetase 1, Biotinylated
- Size:200 μL
- Pack type:Plastic Vial
- Pk:0,1 mg
Specifications
About this item
Western Blot: Approx 80kDa band observed in lysates of cell line A431 (calculated MW of 81.7kDa according to Human NP_002221.1). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: ASS1
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog