Order Entry
Austria
ContactUsLinkComponent
Lead standard solution for ISE calibration, Orion™
Lead standard solution for ISE calibration, Orion™
Catalog # 662-0847
Supplier:  Thermo Orion
CAS Number:  
Lead standard solution for ISE calibration, Orion™
Catalog # 662-0847
Supplier:  Thermo Orion
CAS Number:  

Specifications

  • Concentration:
    0,1M as Pb(ClO₄)₂
  • Pk:
    475 mL
  • Element:
    Lead
  • Symbol:
    Pb

Specifications

About this item