Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Anionic Standards
Iodide Standards
Iodide standard for ISE calibration, Orion™
Iodide standard for ISE calibration, Orion™
Iodide standard for ISE calibration, Orion™
Catalog # 662-0844
Supplier:
Thermo Orion
CAS Number:
Certificates
Expand image
Iodide standard for ISE calibration, Orion™
Catalog # 662-0844
Supplier:
Thermo Orion
CAS Number:
Certificates
Specifications
Concentration:
0,1M
Matrix:
H₂O
Pk:
475 mL
Description:
Iodide standard solution
Specifications
About this item
Product Details
Documentation
Product Details
Documentation