Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Copper Standards
Cupric standard solution for ISE calibration, Orion™
Cupric standard solution for ISE calibration, Orion™
Cupric standard solution for ISE calibration, Orion™
662-0840
:
Thermo Orion
:
Cupric standard solution for ISE calibration, Orion™
662-0840
:
Thermo Orion
:
Concentration:
0,1M
Matrix:
H₂O
Pk:
475 mL
Symbol:
Cu