Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Paper
Thermal paper rolls, SafTest, MP Biomedicals
Thermal paper rolls, SafTest, MP Biomedicals
Thermal paper rolls, SafTest, MP Biomedicals
Catalog # ICNA07MC1030
Supplier:
MP Biomedicals
Certificates
Expand image
Thermal paper rolls, SafTest, MP Biomedicals
Catalog # ICNA07MC1030
Supplier:
MP Biomedicals
Certificates
Specifications
Description:
Thermal paper roll, SafTest, for Microchem I printer, 57,15 mm × 24,4 m
Pk:
1
Specifications
About this item
Product Details
Documentation
Product Details
Documentation