HPLC columns, Viva biphenyl (USP L11)
: Restek
These biphenyl coulmns have a greater degree of dispersion than conventional phenyls and a greater degree of polarisability than phenyl hexyls, creating higher selectivity and a greater range of usability. With heightened interactions shows substantial increases in retention for dipolar, unsaturated, or conjugated solutes and enhanced orthogonal selectivity when using methanol mobile phases.
- Catalog No:
- REST9516553-700
- REST9516532
- REST9516575-700
- REST9516531
- REST9516522-700
- REST9516525
- REST9516573-700
- REST951651E
- REST9516522
- REST9516521
- REST9516515
- REST9516512-700
- REST951650212
- REST9516512
- REST9516575
- REST9516533-700
- REST951650210
- REST9516572
- REST9516532-700
- REST9516555-700
- REST9516572-700
- REST9516525-700
- REST9516552-700
- REST9516565-700
- REST9516523-700
- REST951653E
- REST9516563-700
- REST951657E
- REST9516571
- REST9516565
- REST9516535-700
- REST9516562-700
- REST9516562
- REST951655E
- REST9516561
- REST9516555
- REST9516552
- REST9516551
- REST9516511
- REST9516535
- REST951652E
- REST951656E
- REST9516513-700
- REST9516515-700
- Carbon load:
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
Frequently Bought Together