Order Entry
Austria
ContactUsLinkComponent
Magnesium chloride hexahydrate, crystals Ph. Eur., JPC, USP, FCC, E511, SAFC®
Magnesium chloride hexahydrate, crystals Ph. Eur., JPC, USP, FCC, E511, SAFC®
Catalog # 1.01872.9025
CAS Number:  
Magnesium chloride hexahydrate, crystals Ph. Eur., JPC, USP, FCC, E511, SAFC®
Catalog # 1.01872.9025
CAS Number:  

Specifications

  • Pack type:
    Plastic bag
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :203.3 g/mol
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781