ContactUsLinkComponent
UHPLC columns, Pinnacle® DB PAH UHPLC
UHPLC columns, Pinnacle® DB PAH UHPLC
Supplier:  Restek
Certificates
undefined
UHPLC columns, Pinnacle® DB PAH UHPLC
Supplier:  Restek
Specifications
  • Catalog No:
  • REST9470212
  • REST9470232
  • REST9470252
  • Endcapped:
  • N/A
  • N/A
  • N/A
  • pH range:
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • Temperature limit:
  • 80 °C
  • 80 °C
  • 80 °C