Order Entry
Austria
ContactUsLinkComponent
Cleaning trolley Clino®, Profi C7e
Cleaning trolley Clino®, Profi C7e
Catalog # PPSA2097275
undefined
Cleaning trolley Clino®, Profi C7e
Catalog # PPSA2097275

Specifications

  • Colour:
    Blue
  • Description:
    Trolley with flat wringer and waste disposal 2×70 l, for cleanroom areas
  • W×D×H:
    900×570×1330 mm
  • Pk:
    1

Specifications

About this item

The Clino® Profi C7e trolley system made of stainless steel flat wringer for grey areas.

  • Flat wringer Clino provides defined pressure point for uniform residual moisture
  • Disposal unit can be removed in retirement and is interchangeable and extendable
  • Frame and attachments can be autoclaved
  • Double ball-bearinged guide wheels (125 mm)
Delivery: Supplied with bucket (25 L, 15 L and 3 ×6 L), 3 trays and disposal unit (1 ×70 L).