Anti-POR Rabbit Polyclonal Antibody
Cytochrome p450 is a highly conserved and diverse family of heme-containing proteins that function in the metabolism of numerous endogenous and exogenous substrates.
- Dilution range: Western blot (1:2,000, colorimetric)
- Detects a band of ~78kDa by western blot
Type: Primary
Antigen: POR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
- Catalog No:
- ENZOADIOSA300F
- ENZOADIOSA300D
- Antigen symbol:
- POR
- POR
- Antigen name:
- P450 (cytochrome) oxidoreductase
- P450 (cytochrome) oxidoreductase
- Antigen synonyms:
- CPR|DKFZp686G04235|P450R|CYPOR|FLJ26468
- CPR|DKFZp686G04235|P450R|CYPOR|FLJ26468
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Hamster, Pig, Rabbit, Monkey, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Hamster, Pig, Rabbit, Monkey, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- Native rat Cytochrome p450 Reductase
- Native rat Cytochrome p450 Reductase
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein A affinity chromatography
- Purified by Protein A affinity chromatography
- Storage buffer:
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
- Tested applications:
- Inhibition assay
- Inhibition assay
Frequently Bought Together