Anti-GST-pi Rabbit Polyclonal Antibody
Supplier: Enzo Life Sciences
Certificates
About this item
The GST superfamily of enzymes play an important role in detoxification of xenobiotics by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione.
- Dilution range: Immunohistochemistry (1:500 - 1:1,000), Western blot (1:500 - 1:1,000)
- Detects a band of ~26kDa by western blot
Type: Primary
Antigen: GST-P, Glutathione S-transferase P, GSTP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- ALEXADI-MSA-102-E
- ENZOADIMSA102E
- Antigen symbol:
- GST-pi
- GST-pi
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Human placental GST-pi
- Human placental GST-pi
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein A affinity chromatography
- Purified by Protein A affinity chromatography
- Storage buffer:
- PBS, pH 7,2, containing 50% glycerol
- PBS, pH 7,2, containing 50% glycerol
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together