Order Entry
Austria
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.9%, clear, colourless liquid ACS
Dimethyl sulphoxide ≥99.9%, clear, colourless liquid ACS
Supplier:  MP Biomedicals
Certificates
undefined
Dimethyl sulphoxide ≥99.9%, clear, colourless liquid ACS
Supplier:  MP Biomedicals
Certificates

Specification Test Results

  • Assay:
    ≥99.9%
  • Residue after Evaporation:
    ≤0.01%
  • Titratable Acid:
    ≤0.001 meq/g
  • Water:
    ≤0.1%

Specification Test Results

Related Information

Frequently Bought Together