Order Entry
Austria
ContactUsLinkComponent
Pig alpha-Amylase (from Pancreas), MP Biomedicals
Pig alpha-Amylase (from Pancreas), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Pig alpha-Amylase (from Pancreas), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9000-90-2
  • Source:
    Pancreas
  • Species:
    Pig
  • Storage conditions:
    Store at +4 °C, Do not freeze
  • Enzyme name:
    alpha-Amylase
  • Enzyme activity:
    ≥700 u/mg protein

Specifications

Frequently Bought Together