4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride), yellow powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:28718-90-3
:
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681
:350.25 g/mol
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
Specification Test Results
Frequently Bought Together