Order Entry
Austria
ContactUsLinkComponent
N-Fmoc-L-alanine ≥99.0% (by HPLC), Novabiochem®
N-Fmoc-L-alanine ≥99.0% (by HPLC), Novabiochem®
Certificates
N-Fmoc-L-alanine ≥99.0% (by HPLC), Novabiochem®

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Alternate Names:
    N-α-Fmoc-L-alanine monohydrate
  • Colour (visual):
    white to off white
  • Appearance of substance (visual):
    powder
  • Colour index (0,5 M in DMF):
    ≥60 Hazen
  • Identity (IR):
    passes test
  • Enantiomeric purity:
    ≥99,8% (a/a)
  • Purity (HPLC):
    ≥99,0% (a/a)
  • Fmoc-ß-Ala-OH (HPLC):
    ≤0,1% (a/a)
  • Fmoc-ß-Ala-Ala-OH (HPLC):
    ≤0,1% (a/a)
  • Fmoc-Ala-Ala-OH (HPLC):
    ≤0,1% (a/a)
  • Assay free amino acid (GC):
    ≤0,2%
  • Purity (TLC(011A)):
    ≥98%
  • Purity (TLC(0811)):
    ≥98%
  • Solubility (25 mmole in 50 ml DMF):
    Clearly Soluble
  • Assay (acidimetric):
    ≥93,0%
  • Water (K. F.):
    ≤6,0%
  • Ethyl acetate (HS-GC):
    ≤0,5%
  • Acetate (IC):
    ≤0,02%
  • Storage:
    Store below +30 °C

Specification Test Results

Frequently Bought Together