



Malachite green oxalate, green powder C.I. 42000
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:2437-29-8
:18015-76-4
Formula:C₅₂H₅₄N₄O₁₂
MDL Number:MFCD00011766
:927.02 g/mol
UN:2811
ADR:6.1,III
:GHS05
:GHS07
:GHS08
:GHS09
Specification Test Results
- Identity Test:Passes
- UV/Vis: l max (water) :614 - 618 nm
Specification Test Results
Frequently Bought Together