Order Entry
Austria
ContactUsLinkComponent
Cellulase (from Trichoderma virde), MP Biomedicals
Cellulase (from Trichoderma virde), MP Biomedicals
Catalog # ICNA0215233705
Supplier:  MP Biomedicals
CAS Number:  
Cellulase (from Trichoderma virde), MP Biomedicals
Catalog # ICNA0215233705
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    5 g
  • CAS No.:
    9012-54-8
  • Source:
    Trichoderma virde
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    Store at −0 °C
  • Form:
    Tan-free flowing powder
  • Enzyme name:
    Cellulase
  • Enzyme synonyms:
    1,4-(1,3:1,4)-β-D-Glucan 4-glucano-hydrolase, E.C. 3.2.1.4
  • pH:
    Optimum pH: Varies with the substrate in the range 4,2 - 5,2 (Lit.)

Specifications

About this item

Readily soluble in water.

One unit will liberate one µmole of glucose from cellulose in 1 hr at pH 5 and 37 °C. ; CMC Unit Definition: 100 units is sufficient to produce reducing sugar equivalent to 1 mg of glucose per minute at pH 5,0 and 40 °C.