
1,1-Dimethylbiguanide hydrochloride 98%, white crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1115-70-4
:
Formula:C₄H₁₂ClN₅
MDL Number:MFCD00012582
:165.625 g/mol
:GHS07
Specification Test Results
- Identity Test:Passes
- Purity:#98%
Specification Test Results
Frequently Bought Together