Order Entry
Austria
ContactUsLinkComponent
Lecithin (from soy bean) ≥90%, waxy orange-yellow chunky solid, Water Dispersable Grade
Lecithin (from soy bean) ≥90%, waxy orange-yellow chunky solid, Water Dispersable Grade
Supplier:  MP Biomedicals
Certificates
Lecithin (from soy bean) ≥90%, waxy orange-yellow chunky solid, Water Dispersable Grade
Supplier:  MP Biomedicals
Certificates

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥90%
  • Source:
    Soybeans

Specification Test Results

Related Information

Frequently Bought Together