1-Heptanesulphonic acid sodium salt, hydrate ~98%, white powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate
CAS Number:22767-50-6
:
Formula:H₃C(CH₂)₅CH₂SO₃Na
MDL Number:MFCD00007543
Molecular Weight:202.25 g/mol
Melting Pt:300
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:~98%
- Moisture:7.4 - 9.0%
- FTIR:Conforms to Standard
- Moisture Content:7.4-9.0%
- UV/VIS:(60 mg/ml in water adjusted to pH 3)
OD210 nm:<1.0 - OD220 nm:<0.1
- OD254 nm:<0.04
- OD280 nm:<0.04
Specification Test Results
Frequently Bought Together