Order Entry
Austria
ContactUsLinkComponent
Guanidinium hydrochloride ≥99%, white crystalline powder optical density grade
Guanidinium hydrochloride ≥99%, white crystalline powder optical density grade
Supplier:  MP Biomedicals
Certificates
undefined
Guanidinium hydrochloride ≥99%, white crystalline powder optical density grade
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ~99%
  • FTIR:
    Conforms to Standard
  • Melting Point:
    180 - 190 °C
  • OD260 (6,0 M aq soln):
    ≤0,03
  • Solubility (6,0 M aq soln) C:
    Clear, Colorless Solution

Specification Test Results

Frequently Bought Together