Order Entry
Austria
ContactUsLinkComponent
Ammonium iron(II) sulphate hexahydrate 98.5-101.5%, crystalline ACS
Irritant
Ammonium iron(II) sulphate hexahydrate 98.5-101.5%, crystalline ACS
Supplier:  MP Biomedicals
Certificates
Irritant
Ammonium iron(II) sulphate hexahydrate 98.5-101.5%, crystalline ACS
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Assay:
    98,5 - 101,5%
  • Insoluble Matter:
    ≤0,01%
  • Phosphate (PO4):
    <0,003%
  • Copper (Cu):
    <0,003%
  • Manganese (Mn):
    <0,01%
  • Zinc (Zn):
    0,003%
  • Ferric Ion (Fe3+):
    0,01%

Specification Test Results

Frequently Bought Together