
Ammonium heptamolybdate tetrahydrate, white crystalline powder ACS
:12054-85-2
:12027-67-7
:(NH₄)₆Mo₇O₂₄·4H₂O
:MFCD00167059
:1235.86 g/mol
:GHS07
:Ambient
- Assay:(as MoO3) 81,0 - 83,0%
- Heavy Metals:(as Pb) ≤0,001%
- Insoluble Matter:≤0,005%
- Chloride (Cl):≤0,002%
- Arsenate, Phosphate and Silicate (as SiO2):≤0,001%
- Phosphate (PO4):≤5 ppm
- Sulphate (SO4):≤0,02%
- Magnesium:≤,005%
- Potassium:≤0,002%
- Sodium:≤0,01%
Frequently Bought Together