Order Entry
Austria
ContactUsLinkComponent
Poly-L-lysine hydrobromide, MP Biomedicals
Poly-L-lysine hydrobromide, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Poly-L-lysine hydrobromide, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    25988-63-0
  • Storage conditions:
    Freezer
  • Form:
    Beige Powder
  • MDL No.:
    MFCD00237230
  • Protein/peptide name:
    Poly-L-lysine hydrobromide
  • Molecular weight:
    30,000 - 70,000 kDa
  • Molecular formula:
    (C₆H₁₃N₂OBr)n

Specifications

Frequently Bought Together