Sulphanilamide, white powder
ICNA0210298890
:
- Pk:500 g
- Seq:3645
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:160 - 170°C
- UV/VIS: λmax (water):255 - 259 nm
:
:172.21 g/mol
:160…170 °C
:1.08 g/mL (lit.)
:Ambient
:63-74-1
:H₂NC₆H₄SO₂NH₂
:MFCD00007939