Order Entry
Austria
ContactUsLinkComponent
Dextran sulphate sodium salt, M.W. (6,000-8,000), white powder
Dextran sulphate sodium salt, M.W. (6,000-8,000), white powder
Supplier:  MP Biomedicals
Certificates
Dextran sulphate sodium salt, M.W. (6,000-8,000), white powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • pH (1% aq soln):
    5.0 - 7.5
  • Specific Rotation :
    +75 ° to +105 °
  • Loss on Drying:
    10%
  • Residue on Ignition:
    35 - 50%
  • Molecular weight:
    Passes

Specification Test Results

Frequently Bought Together