Order Entry
Austria
ContactUsLinkComponent
Human FGF4 ELISA kit
Human FGF4 ELISA kit
Catalog # BIRBORB138053-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human FGF4 ELISA kit
Catalog # BIRBORB138053-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human FGF4 ELISA Kit
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human FGF4 ELISA Kit for the quantitative detection of human fibroblast growth factor 4 (FGF-4) in cell culture supernates, serum and plasma (heparin, EDTA).

FGF-4 is a protein that in humans is encoded by the FGF4 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumour growth and invasion.