Order Entry
Austria
ContactUsLinkComponent
Mouse PTX3 ELISA kit
Mouse PTX3 ELISA kit
Catalog # BIRBORB96797-96
Supplier:  Biorbyt
CAS Number:  
undefined
Mouse PTX3 ELISA kit
Catalog # BIRBORB96797-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    PTX3
  • Description:
    Mouse PTX3 ELISA Kit
  • Specificity:
    Natural and recombinant mouse PTX3
  • Detection range:
    156 - 10000 pg/ml
  • Sensitivity:
    <10pg/ml

Specifications

About this item

Mouse PTX3 ELISA Kit for the quantitative detection of mouse pentraxin 3.

  • No detectable cross-reactivity with other relevant proteins

Pentraxin 3 is a member of the pentraxin superfamily. This super family is characterised by cyclic multimeric structure. The PTX3 gene is mapped to 3q25.32. The predicted 381-amino acid PTX3 protein has homology to the pentraxin protein family. Significant levels of PTX3 were detected in plasma of neutropenic patients with systemic A. PTX3 is effective in preventing CMV infection and reactivation, as well as subsequent Aspergillus infection. PTX3 activates the classical pathway of complement activation and facilitates pathogen recognition by macrophages and DCs.