Order Entry
Austria
ContactUsLinkComponent
 
Mouse CSF1R ELISA kit
Catalog # BIRBORB96791-96
Supplier:  Biorbyt
CAS Number:  
undefined
Mouse CSF1R ELISA kit
Catalog # BIRBORB96791-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    CSF1R
  • Description:
    Mouse CSF1R ELISA Kit
  • Specificity:
    Natural and recombinant mouse CSF1R
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Mouse CSF1R ELISA Kit is a sandwich ELISA kit for the quantitative detection for mouse CSF1R (Colony Stimulating Factor 1 Receptor, M-CSFR, CD115). CSF1R is a cell-surface protein encoded, in humans, by the CSF1R gene.

  • No detectable cross-reactivity with other relevant proteins

The CSF1R gene is located on the long arm of chromosome 5 5q32 on the Crick minus strand. The encoded protein is a single pass type I membrane protein and acts as the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages.