ContactUsLinkComponent
Rat CRP/C reactive protein ELISA kit
Rat CRP/C reactive protein ELISA kit
Catalog # BIRBORB79257-96
Supplier:  Biorbyt
CAS Number:  
Rat CRP/C reactive protein ELISA kit
Catalog # BIRBORB79257-96
Supplier:  Biorbyt
CAS Number:  
Specifications
  • Pk:
    96
  • Primary antibody reactivity:
    Rat
  • Description:
    Rat C Reactive Protein ELISA Kit
Specifications
About this item
Rat C Reactive Protein (CRP) ELISA Kit is used for the quantitative determination of rat C reactive protein in serum, plasma or cell culture supernatant.