Order Entry
Austria
ContactUsLinkComponent
Human Osteoactivin ELISA kit
Human Osteoactivin ELISA kit
Catalog # BIRBORB79180-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human Osteoactivin ELISA kit
Catalog # BIRBORB79180-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human Osteoactivin ELISA Kit
  • Detection range:
    45 - 12000 pg/ml
  • Sensitivity:
    45 pg/ml

Specifications

About this item

Human Osteoactivin ELISA Kit for the quantitative measurement of human osteoactivin (GPNMB) in serum, plasma, cell culture supernatants and urine.

Delivery: Kit includes 96-well microplate precoated with anti-human osteoactivin, wash buffer concentrate 20X (25 ml), standards (2 vials), assay diluent A (30 ml), assay diluent B 5X (15 ml), detection antibody osteoactivin (2 vial), HRP-streptavidin concentrate 500X (200 µl), TMB one-step substrate reagent (12 ml), and stop solution (8 ml).