ContactUsLinkComponent
Adenosine 5'-monophosphate monohydrate free acid (from yeast), white powder
Adenosine 5'-monophosphate monohydrate free acid (from yeast), white powder
Supplier:  MP Biomedicals
Certificates
Adenosine 5'-monophosphate monohydrate free acid (from yeast), white powder
Supplier:  MP Biomedicals
Specification Test Results
  • Identity Test:
    Passes
  • Source:
    Yeast
  • FTIR:
    Conforms to Standard
  • UV/Vis: λmax (water):
    258 ± 5 n
  • OD250/OD260:
    0.83 ± 0.0
  • OD 280/OD26:
    0.15 ± 0.0
  • Loss on Drying:
    <7%