ContactUsLinkComponent
Labels for BMP™ 51/53 printer, type B-595
Labels for BMP™ 51/53 printer, type B-595
Catalog # BRDY170949
Supplier:  Brady
undefined
Labels for BMP™ 51/53 printer, type B-595
Catalog # BRDY170949
Supplier:  Brady
Specifications
  • Type:
    B-595
  • Colour:
    Black/white
  • Dimensions:
    6,35 mm×7,62 m
  • Material:
    Vinyl
  • Seq:
    40
  • Model:
    M4C-250-595-WT-BK
  • Packed:
    1 label/box
  • Pk:
    1
  • Adhesive:
    Permanent acrylic
Specifications
About this item
These are made from durable, low-shrink vinyl with a very aggressive adhesive. The labels adhere to irregular, curved, rough and highly textured surfaces (painted cinder blocks, uneven wood, textured plastics, paper jacketed pipes, powder coated surfaces). Also adhere to difficult low energy surface items (PVC piping, blow-moulded equipment cases, ABS plastics, recycled plastics).