ContactUsLinkComponent
Vector, pUC 18, MP Biomedicals
Vector, pUC 18, MP Biomedicals
Catalog # ICNA04821226
Supplier:  MP Biomedicals
CAS Number:  
Vector, pUC 18, MP Biomedicals
Catalog # ICNA04821226
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    25 µG
  • Description:
    Vector, pUC 18
Specifications
About this item
pUC 18 is a small vector of approximately 2,7 Kb containing the Pvu II/EcoR I fragment of pBR322 and the lacZ gene multiple cloning site from M13mp18.