Orders LinkContactUsLinkComponent
Temperature probes
Temperature probes
Supplier:   Industrial Physics
Temperature probes
Supplier:  Industrial Physics
Certificates
Temperature probes
Temperature probes
6 Options Available Below
Product Details & Documents
These temperature probes can be connected to instruments suited for changeable probes (type ‘K’) using a plug connection.
  • Provided with a firm hand-grip and a one meter curled cable
  • Thicker probes have a longer response time
Documents
Product Family Options
Product Information
  • Type
    Range
    Ø×L
    Availability
    Price
  • SIMETE5103
    SquaNon- contact surface, MX-series model MX2G-MX4PGre surface probe
    SquaNon- contact surface, MX-series model MX2G-MX4PGre surface probe
    Type 80111
    0…200 °C
    8×130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 365,00 €
    365,00 €
    1 * 1 ST
       365,00 €
  • SIMETE5104
    Liquid probe
    Liquid probe
    Type 80104
    0…1100 °C
    3×130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 187,00 €
    187,00 €
    1 * 1 ST
       187,00 €
  • SIMETE5105
    Liquid probe
    Liquid probe
    Type 80105
    0…900 °C
    1,5×130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 165,00 €
    165,00 €
    1 * 1 ST
       165,00 €
  • SIMETE5106
    Needle probe
    Needle probe
    Type 80106
    0…600 °C
    3×130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 148,00 €
    148,00 €
    1 * 1 ST
       148,00 €
  • SIMETE5110
    Surface probe
    Surface probe
    Type 80110
    0…900 °C
    8×130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 88,60 €
    88,60 €
    1 * 1 ST
       88,60 €
  • SIMETE5125
    Heavy Duty asphalt probe with sturdy T-shaped hand-grip
    Heavy Duty asphalt probe with sturdy T-shaped hand-grip
    Type 133-130 ETI
    0…300 °C
    8×500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €