- Description:Linear abraser, Taber® 5750
- Pk:1
This linear wear resistance tester is designed to test virtually any size or shape specimen. Ideal for testing material properties of contoured surfaces and finished products.
- Eleven stroke lengths (0,2" to 4,0")
- Variable stroke speed from 2 to 75 cycles per minute
- Preset stroke speed buttons for 2, 15, 25, 30, 40, 60 cycles per minute
- Variable load from 350 to 2100 g with optional weight discs
- Stainless Steel Wearaser Collet for use with vitrified or resilient Wearsers
- Laser alignment guide
- Programmable up to 999 999 cycles
The linear abraser uses a free-floating head to follow the contours of every sample, permitting testing of finished products. With virtually no limit on sample size or shape, the Linear Abraser is ideal for testing consumer products, automotive components, painted parts, printed graphics, optical products, rubber, leather, textiles, and for use in testing laboratories.
With adjustable stroke length, speed, load, and a wide selection of Taber attachments, the Linear Abraser allows you to customise test parameters to your exact needs. Optional attachments can be used to perform scratch, coin scrape, Crockmeter, rub, pencil hardness scratch, paperclip mar, magnetic stripe abrasion, wire insulation scrape, chemical rub, plus custom testing.
The linear abraser abrades with a Wearaser®. The size and shape of a pencil eraser, the Wearaser uses the same high-quality Taber abrasive media found in our world-famous wheels so you're assured of consistent test results.