Orders LinkContactUsLinkComponent
Agarose, MetaPhor™
Agarose, MetaPhor™
Supplier:   LONZA
Agarose, MetaPhor™
Supplier:  LONZA
Certificates
Agarose, MetaPhor™
Agarose, MetaPhor™
3 Options Available Below
Product Details & Documents
Intermediate melting point agarose ideal for resolution of DNA fragments, PCR and RT-PCR products, differing in size by 2%, in the range of 20 to 800 bp, by submarine gel electrophoresis.
MetaPhor™ agarose gel is ideal for resolving AMPFLPs, STRs, and tri- and tetranucleotide repeats.
Product Family Options
Product Information
  • Availability
    Price
  • 733-1211
    Agarose, MetaPhor™
    Agarose, MetaPhor™
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 304,00 €
    304,00 €
    1 * 25 g
       304,00 €
  • 733-1210
    Agarose, MetaPhor™
    Agarose, MetaPhor™
    Log in for availability
    Log in for availability
    1 * 125 g
    1 * 125 g - 1 020,00 €
    1 020,00 €
    1 * 125 g
       1 020,00 €
  • 733-1212
    Agarose, MetaPhor™
    Agarose, MetaPhor™
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 3 240,00 €
    3 240,00 €
    1 * 500 g
       3 240,00 €