ContactUsLinkComponent
Wood and plastic moisture tester, Skipper Plus
Wood and plastic moisture tester, Skipper Plus
Catalog # SIMELI9012
Supplier:  Industrial Physics
undefined
Wood and plastic moisture tester, Skipper Plus
Catalog # SIMELI9012
Supplier:  Industrial Physics
Specifications
  • Model:
    Skipper Plus
  • Description:
    Wood and plastic moisture tester
  • Pk:
    1
Specifications
About this item
Skipper Plus is a non-destructive moisture detector designed for locating damp in wood or laminated plastics such as polyester.
  • Short rubber electrodes, which do not damage the surface
  • Three moisture ranges for moisture in both fresh and salt water
  • Developed for osmosis-control and measuring moisture before painting yachts
W×D×H: 150×80×30 mm