Surface probes, clamp-type, for Curvex datalogger
Supplier: Industrial Physics
Certificates
About this item
Small and elegant clamp-type probes to measure temperature of any object.
- Max. object thickness is 20 mm
- Energy and/or water efficient
- Recycled/renewable content
Silver tipped sensor is thermally isolated from the clamp by ceramics. The capton-wrapped, metal braid and PTFE coated probe cables last significantly longer than other designs.
Frequently Bought Together