Adapters, tubing, KA, KECK™, DURAN®
KECK™ adapters are made up of glass-fibre reinforced, thermoplastic polyester (PBT) for glass hose couplings KA.
- Protect fingers and hands from being injured
- Prevents the breakage of glass hose couplings
- Connect or remove hose by twisting ¼ turn
- The clip, O-ring and the screwon cap are screwed onto the hose coupling permanently
Hose can be connected quickly, safely and without leakage (upto 30 bar pressure, 0,66 mbar vacuum).
: The adapters are available as a complete set which comes either with 4,5 / 9,0 mm Ø tubing part.
Frequently Bought Together