PIG Blue® absorbent mat pads and rolls
Lieferant: NEW PIG
Zertifikate
Über diesen Artikel
PIG Blue is the most absorbent mat. Maximum absorbency soaks up more so you use less on big spills and high-volume leaks.
- Consistent construction produces a mat that's free of light spots to absorb liquid evenly for better wicking and retention
- FibreFusion Technology fuses superabsorbent natural fibres with strong synthetic fibres for one high-performance absorbent
- Stays together, even when picking up or shifting a fully saturated mat
- Contains 90% recycled fibres; ideal in situations where green products are desired or required
- Fire retardant for added safety; flame-tested in accordance with ASTM Standard E84-08
- Absorbs oils, coolants, solvents and water; not recommended for acids, caustics or other corrosive liquids
Rolls are ideal for covering large areas and soaking up leaks and overspray. Easy-tear perforations let you take only what you need, so you use less mat and save money. Heavy-weight Pig Blue construction is super absorbent for your most demanding tasks and large-volume cleanups.
Frequently Bought Together