Order Entry
Austria
ContactUsLinkComponent
Multi-gas detector, GasAlert MicroClip
Multi-gas detector, GasAlert MicroClip
Catalog # SIMEDM6000
Supplier:  Industrial Physics
undefined
Multi-gas detector, GasAlert MicroClip
Catalog # SIMEDM6000
Supplier:  Industrial Physics

Specifications

  • Model:
    MicroClip
  • Parameter(s):
    O₂, % LEL
  • Temperature range:
    −20…+50 °C
  • W×D×H:
    107×60×27 mm
  • Battery life:
    12 hours
  • Alarm:
    Audible, visual, Buzzer in the event of low, high, TWA and STEL conditions
  • Weight:
    160 g
  • Battery type:
    Rechargeable Lithium
  • Pk:
    1

Specifications

About this item

The GasAlert MicroClip offers simultaneous monitoring of two gasses (O₂, LEL) in a rugged compact case for confined areas.

  • Hazardous materials (HAZMAT) response and other applications requiring gas monitoring
  • One-button operation with automatic calibration
  • Can record and transmit up to ten gas alarm events
Delivery: Supplied with sensors, lithium battery, stainless steel belt clip, calibration hose/cap and manual.